Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-phenylglycine tert-butyl ester hydrochloride | 256478-95-2 | MFCD03093480 | >97.0% | 243.73 g/mol | C12H18ClNO2 | 100 G
SDP

Catalog No. 5000209427
Encompass_Preferred

(R)-Phenylglycine tert-butyl ester hydrochloride is the R-enantiomer of phenylglycine tert-butyl ester in its hydrochloride salt form, supplied as a white to off-white solid for research and synthetic uses.

  • Chiral R-enantiomer for stereoselective synthesis.
  • Hydrochloride salt for improved stability and handling.
  • White to off-white solid physical form.
  • Molecular weight 243.73 g/mol.
  • Chemical formula C12H18ClNO2.
  • Available in common laboratory pack sizes, including 100 g.

Catalog No. 50-002-09427 Supplier Medchemexpress LLC Supplier No. HYW042009A100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-Phenylglycine tert-butyl ester hydrochloride is the R-enantiomer of phenylglycine tert-butyl ester in its hydrochloride salt form, supplied as a white to off-white solid for research and synthetic uses.

  • Chiral R-enantiomer for stereoselective synthesis.
  • Hydrochloride salt for improved stability and handling.
  • White to off-white solid physical form.
  • Molecular weight 243.73 g/mol.
  • Chemical formula C12H18ClNO2.
  • Available in common laboratory pack sizes, including 100 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.