Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ R-Phycoerythrin

Catalog No. P801
Encompass_Preferred
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P801 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P801 Supplier Invitrogen™ Supplier No. P801
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

R-Phycoerythrin

Phycobiliprotein bioconjugates including antibody and other protein conjugates can be made with the exceptionally bright phycobiliprotein, R-phycoerythrin using the Protein-Protein Crosslinking Kit (P6305).
TRUSTED_SUSTAINABILITY

Specifications

Concentration 4 mg/mL
Quantity 0.5 mL
Product Type R-Phycoerythrin
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.