Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ R-Phycoerythrin, Biotin-XX Conjugate

Catalog No. P811
Encompass_Preferred
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P811 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P811 Supplier Invitrogen™ Supplier No. P811
Add to Cart
Edge
Add to Cart

R-Phycoerythrin, Biotin-XX Conjugate

R-phycoerythrin biotin-XX conjugate consists of the exceptionally bright phycobiliprotein, R-PE conjugated with biotin-XX, where the biotin is well separated from the point of attachment by a fourteen-atom spacer.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 4 mg/mL
Conjugate Biotin
Quantity 0.5 mL
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Shipping Condition Wet Ice
Form Liquid
Protein Family Other Proteins
Product Type Biotin Conjugate (R-PE)
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.