Learn More
Dojindo Molecular Technologies Inc R-Phycoerythrin Labeling Kit-NH2 (3 samples) is used to prepare R-Phycoerythrin-labeled IgG, high recovery of conjugates in 3 hours, all processes in a single filtration tube, 50-200 µg/sample.
SDP
Catalog No. NC1960988
Encompass_Preferred
R-Phycoerythrin (R-PE) is one of the phycobiliproteins. It emits a red fluorescence at around 578 nm and can be excited at 488 nm. Because of this high fluorescence, phycobiliprotein-labeled antibodies/molecules can provide greater sensitivity in flow cytometry and immunostaining. R-phycoerythrin Labeling Kit-NH2 is designed for the preparation of R-PE-labeled IgG. This kit contains all of the reagents necessary for R-PE labeling, including the Storage buffer for conjugates. NH2-reactive R-PE (a component of this kit) has an activated ester group and can easily make a covalent bond with an amino group of the target molecule without any activation process. The filtration tube in this kit allows a quick buffer exchange and concentration of sample IgG solution. Dojindo offers various protein labeling reagents. (https://www.dojindo.com/labeling-chemistry/)
Catalog No. NC1960988 Supplier Dojindo Molecular Technologies Inc Supplier No. LK2310
R-Phycoerythrin (R-PE) is one of the phycobiliproteins. It emits a red fluorescence at around 578 nm and can be excited at 488 nm. Because of this high fluorescence, phycobiliprotein-labeled antibodies/molecules can provide greater sensitivity in flow cytometry and immunostaining. R-phycoerythrin Labeling Kit-NH2 is designed for the preparation of R-PE-labeled IgG. This kit contains all of the reagents necessary for R-PE labeling, including the Storage buffer for conjugates. NH2-reactive R-PE (a component of this kit) has an activated ester group and can easily make a covalent bond with an amino group of the target molecule without any activation process. The filtration tube in this kit allows a quick buffer exchange and concentration of sample IgG solution. Dojindo offers various protein labeling reagents. (https://www.dojindo.com/labeling-chemistry/)