Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ R-Phycoerythrin, Pyridyldisulfide Derivative

Catalog No. P806
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P806 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P806 Supplier Invitrogen™ Supplier No. P806
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

R-Phycoerythrin, Pyridyldisulfide Derivative

The pyridyldisulfide derivative of R-phycoerythrin (R-PE) can be reacted directly with thiolated antibodies, enzymes and other biomolecules to form a disulfide linkage.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 mg/mL
Quantity 1 mL
Product Type Pyridyldisulfide Derivative of R-phycoerythrin
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Shipping Condition Wet Ice
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.