Learn More
Medchemexpress LLC (R)-pirtobrutinib | 2101700-14-3 | 99.8% | C22H21F4N5O3 | 10MG
SDP
Catalog No. 5000260255
Encompass_Preferred
(R)-Pirtobrutinib is the R-enantiomer of pirtobrutinib (LOXO-305), a selective, non-covalent Bruton's tyrosine kinase (BTK) inhibitor used for biochemical and cellular research into BTK signaling and resistance mutations, including C481 substitutions. The compound is supplied as a high-purity solid with established solubility and storage profiles for laboratory use.
  • High purity (99.8%).
  • Selective, non-covalent BTK inhibitor active against C481 substitution mutations.
  • Soluble in DMSO (≈200 mg/mL); ultrasonic assistance recommended.
  • Light yellow to yellow solid appearance.
  • Stable under defined storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.
Catalog No. 50-002-60255 Supplier Medchemexpress LLC Supplier No. HY131328A10MG
(R)-Pirtobrutinib is the R-enantiomer of pirtobrutinib (LOXO-305), a selective, non-covalent Bruton's tyrosine kinase (BTK) inhibitor used for biochemical and cellular research into BTK signaling and resistance mutations, including C481 substitutions. The compound is supplied as a high-purity solid with established solubility and storage profiles for laboratory use.
  • High purity (99.8%).
  • Selective, non-covalent BTK inhibitor active against C481 substitution mutations.
  • Soluble in DMSO (≈200 mg/mL); ultrasonic assistance recommended.
  • Light yellow to yellow solid appearance.
  • Stable under defined storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.