Learn More
Meso Scale Discovery R-PLEX AB38 4G8 ASY
SDP
Catalog No. 5000078813
Encompass
The Meso Scale Discovery R-PLEX Human Abeta38 (4G8) Assay Kit is a multiplex immunoassay that measures Abeta38 in human serum plasma CSF and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a plate coated with a 4G8 antibody to capture Abeta38 It also includes detection antibodies standards buffers calibrators controls and substrate The kit is for research use only and not for use in diagnostic procedures It allows sensitive accurate and reproducible quantification of Abeta38 levels
Catalog No. 50-000-78813 Supplier Meso Scale Discovery Supplier No. K1509NR2
The Meso Scale Discovery R-PLEX Human Abeta38 (4G8) Assay Kit is a multiplex immunoassay that measures Abeta38 in human serum plasma CSF and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a plate coated with a 4G8 antibody to capture Abeta38 It also includes detection antibodies standards buffers calibrators controls and substrate The kit is for research use only and not for use in diagnostic procedures It allows sensitive accurate and reproducible quantification of Abeta38 levels