Learn More
Meso Scale Discovery R-PLEX AB38 4G8 ASYQP
SDP
Catalog No. 5000080524
Encompass
The Meso Scale Discovery R-PLEX Human Abeta38 (4G8) Assay Kit is a multiplex immunoassay that quantitatively measures Abeta38 in human serum plasma CSF and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a plate coated with capture antibody detection antibody read buffer sample diluent calibrator diluent wash buffer and calibrators to generate a standard curve It allows rapid and accurate measurement of Abeta38 levels
Catalog No. 50-000-80524 Supplier Meso Scale Discovery Supplier No. K1509NR22
The Meso Scale Discovery R-PLEX Human Abeta38 (4G8) Assay Kit is a multiplex immunoassay that quantitatively measures Abeta38 in human serum plasma CSF and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a plate coated with capture antibody detection antibody read buffer sample diluent calibrator diluent wash buffer and calibrators to generate a standard curve It allows rapid and accurate measurement of Abeta38 levels