Learn More
Meso Scale Discovery R-PLEX AB40 4G8 ASY
SDP
Catalog No. 5000081776
Encompass
The Meso Scale Discovery R-PLEX Human Abeta40 4G8 Assay Kit is a multiplex immunoassay that measures Abeta40 levels in human samples such as serum plasma CSF and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a plate coated with a 4G8 antibody to capture Abeta40 along with detection antibodies calibrators controls buffers and other reagents needed to run the assay It allows rapid and accurate quantification of Abeta40 levels which may be useful as a biomarker in Alzheimer s disease research and drug development
Catalog No. 50-000-81776 Supplier Meso Scale Discovery Supplier No. K1509LR2
The Meso Scale Discovery R-PLEX Human Abeta40 4G8 Assay Kit is a multiplex immunoassay that measures Abeta40 levels in human samples such as serum plasma CSF and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a plate coated with a 4G8 antibody to capture Abeta40 along with detection antibodies calibrators controls buffers and other reagents needed to run the assay It allows rapid and accurate quantification of Abeta40 levels which may be useful as a biomarker in Alzheimer s disease research and drug development