Learn More
Meso Scale Discovery R-PLEX AB40 4G8 ASYQP
SDP
Catalog No. 5000079229
Encompass
The R-PLEX Human Abeta40 (4G8) Assay Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures Abeta40 in human serum plasma CSF and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibody detection antibody read buffer sample diluent calibrators and controls It can detect Abeta40 levels as low as 1 95 pg/mL with an approximate 3 5 hour total assay time
Catalog No. 50-000-79229 Supplier Meso Scale Discovery Supplier No. K1509LR22
The R-PLEX Human Abeta40 (4G8) Assay Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures Abeta40 in human serum plasma CSF and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibody detection antibody read buffer sample diluent calibrators and controls It can detect Abeta40 levels as low as 1 95 pg/mL with an approximate 3 5 hour total assay time