Learn More
Meso Scale Discovery R-PLEX AB42 4G8 ASY
SDP
Catalog No. 5000081827
Encompass
The Meso Scale Discovery R-PLEX Human Abeta40 6E10 Assay is a multiplex immunoassay kit for detecting amyloid beta 40 (Abeta40) and amyloid beta bound by antibody 6E10 in human samples like serum plasma and CSF It uses electrochemiluminescence (ECL) technology for high sensitivity and a broad dynamic range The assay requires 50 L or less of sample and can be completed in 3 5 hours It includes a 96-well plate with capture antibodies detection antibodies standards calibrators controls diluent read buffer wash buffer and plate seals
Catalog No. 50-000-81827 Supplier Meso Scale Discovery Supplier No. K1509MR2
The Meso Scale Discovery R-PLEX Human Abeta40 6E10 Assay is a multiplex immunoassay kit for detecting amyloid beta 40 (Abeta40) and amyloid beta bound by antibody 6E10 in human samples like serum plasma and CSF It uses electrochemiluminescence (ECL) technology for high sensitivity and a broad dynamic range The assay requires 50 L or less of sample and can be completed in 3 5 hours It includes a 96-well plate with capture antibodies detection antibodies standards calibrators controls diluent read buffer wash buffer and plate seals