Learn More
Meso Scale Discovery R-PLEX AB42 4G8 ASYQP
SDP
Catalog No. 5000081828
Encompass
The Meso Scale Discovery R-Plex Human Abeta40 6E10 Assay is a multiplex immunoassay kit that allows simultaneous measurement of amyloid beta 40 (Abeta40) and total amyloid beta (6E10) in human samples such as CSF serum and plasma It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay requires an MSD instrument such as SECTOR Imager 2400 or 6000 for analysis It can aid in understanding amyloid beta levels related to Alzheimer s disease research The kit includes a plate coated with capture antibodies detection antibodies calibrator and diluent
Catalog No. 50-000-81828 Supplier Meso Scale Discovery Supplier No. K1509MR22
The Meso Scale Discovery R-Plex Human Abeta40 6E10 Assay is a multiplex immunoassay kit that allows simultaneous measurement of amyloid beta 40 (Abeta40) and total amyloid beta (6E10) in human samples such as CSF serum and plasma It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay requires an MSD instrument such as SECTOR Imager 2400 or 6000 for analysis It can aid in understanding amyloid beta levels related to Alzheimer s disease research The kit includes a plate coated with capture antibodies detection antibodies calibrator and diluent