Learn More
Meso Scale Discovery R-PLEX Human CEACAM-5 Antibody Set, 5 plate size
SDP
Catalog No. NC3236795
Encompass
The R-PLEX platform offers matched sets of antibodies and calibrators for a wide menu of biomarkers. The R-PLEX Human CEACAM-5 Antibody Set can be used with other MSD components (see "Recommended Components" tab below) to build assays that measure CEACAM-5. These assays can measure CEACAM-5 alone or in multiplex panels with other biomarkers.
Catalog No. NC3236795 Supplier Meso Scale Discovery Supplier No. F21QE3
The R-PLEX platform offers matched sets of antibodies and calibrators for a wide menu of biomarkers. The R-PLEX Human CEACAM-5 Antibody Set can be used with other MSD components (see "Recommended Components" tab below) to build assays that measure CEACAM-5. These assays can measure CEACAM-5 alone or in multiplex panels with other biomarkers.