Learn More
Meso Scale Discovery R-PLEX Human p21 Total Assay, Singleplex, 5 plates, 96-well, Sensitivity 13.9 pg/mL, Dynamic Range 13.9-70,000 pg/mL, Sample Types: Plasma, Serum
SDP
Catalog No. NC3353458
Encompass
R-PLEX Human p21 Total Assay, Singleplex, 5 plates, 96-well, Sensitivity 13.9 pg/mL, Dynamic Range 13.9-70,000 pg/mL, Sample Types: Plasma, Serum
Catalog No. NC3353458 Supplier Meso Scale Discovery Supplier No. K151ARMR2
R-PLEX Human p21 Total Assay, Singleplex, 5 plates, 96-well, Sensitivity 13.9 pg/mL, Dynamic Range 13.9-70,000 pg/mL, Sample Types: Plasma, Serum