Learn More
Meso Scale Discovery R-PLEX Human 4-1BB/TNFRSF9 ASY
SDP
Catalog No. 5000078889
Encompass
The R-PLEX Human 4-1BB/TNFRSF9 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human 4-1BB/TNFRSF9 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for detecting multiple protein biomarkers on one plate The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are available in 3 5 hours with an MSD instrument offering high sensitivity a wide dynamic range and requiring only 25-50 L of sample
Catalog No. 50-000-78889 Supplier Meso Scale Discovery Supplier No. K1518UR2
The R-PLEX Human 4-1BB/TNFRSF9 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human 4-1BB/TNFRSF9 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for detecting multiple protein biomarkers on one plate The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are available in 3 5 hours with an MSD instrument offering high sensitivity a wide dynamic range and requiring only 25-50 L of sample