Learn More
Meso Scale Discovery R-PLEX Human 4-1BBL/TNFSF9 ASY
SDP
Catalog No. 5000078336
Encompass
The R-PLEX Human 4-1BBL (TNFSF9) Assay is a multiplex immunoassay kit by Meso Scale Diagnostics that allows for the quantitative measurement of human 4-1BBL in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 80 tests across two plates It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels and calibrator diluents to generate a standard curve
Catalog No. 50-000-78336 Supplier Meso Scale Discovery Supplier No. K151ALLR2
The R-PLEX Human 4-1BBL (TNFSF9) Assay is a multiplex immunoassay kit by Meso Scale Diagnostics that allows for the quantitative measurement of human 4-1BBL in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 80 tests across two plates It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels and calibrator diluents to generate a standard curve