Learn More
Meso Scale Discovery R-PLEX Human AB total ASY
SDP
Catalog No. 5000081717
Encompass
The Meso Scale Discovery R-PLEX Human Abeta Total Assay is an electrochemiluminescent immunoassay for quantifying total Abeta in human serum plasma and CSF It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible Abeta measurement The assay quantifies from 1 22 to 1000 pg/mL without cross-reactivity with other amyloid proteins It requires 50 L of sample and is compatible with MSD s SECTOR Imager 6000 for high throughput The R-PLEX Abeta Total Assay is a precise tool for Alzheimer s disease research
Catalog No. 50-000-81717 Supplier Meso Scale Discovery Supplier No. K1518HR2
The Meso Scale Discovery R-PLEX Human Abeta Total Assay is an electrochemiluminescent immunoassay for quantifying total Abeta in human serum plasma and CSF It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible Abeta measurement The assay quantifies from 1 22 to 1000 pg/mL without cross-reactivity with other amyloid proteins It requires 50 L of sample and is compatible with MSD s SECTOR Imager 6000 for high throughput The R-PLEX Abeta Total Assay is a precise tool for Alzheimer s disease research