Learn More
Meso Scale Discovery R-PLEX Human AB total ASYQP
SDP
Catalog No. 5000080158
Encompass
The Meso Scale Discovery K1518HR-22 is a 96-well electrochemiluminescent immunoassay plate for the quantitative detection of human IL-15 It utilizes MSD s patented SULFO-TAG labels and carbon electrodes to enable high sensitivity and broad dynamic range The plate is pre-coated with a capture antibody specific for human IL-15 The limit of detection is 1 5 pg/mL and the upper limit of quantitation is 10 000 pg/mL The assay takes approximately 3 5 hours to complete This product allows for the accurate and sensitive measurement of IL-15 levels in serum plasma cell culture supernatants and other biological samples
Catalog No. 50-000-80158 Supplier Meso Scale Discovery Supplier No. K1518HR22
The Meso Scale Discovery K1518HR-22 is a 96-well electrochemiluminescent immunoassay plate for the quantitative detection of human IL-15 It utilizes MSD s patented SULFO-TAG labels and carbon electrodes to enable high sensitivity and broad dynamic range The plate is pre-coated with a capture antibody specific for human IL-15 The limit of detection is 1 5 pg/mL and the upper limit of quantitation is 10 000 pg/mL The assay takes approximately 3 5 hours to complete This product allows for the accurate and sensitive measurement of IL-15 levels in serum plasma cell culture supernatants and other biological samples