Learn More
Meso Scale Discovery R-PLEX Human AB38 6E10 ASY
SDP
Catalog No. 5000079029
Encompass
The Meso Scale Discovery Human Abeta38 6E10 Kit is a multiplex immunoassay that quantitatively measures amyloid beta 38 (Abeta38) in human samples such as serum plasma CSF and cell culture supernatants This electrochemiluminescence (ECL) assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrator and sample diluent The 6E10 antibody enables specific detection of Abeta38 This assay allows high-throughput quantification of Abeta38 levels which may be useful as a biomarker in Alzheimer s disease research
Catalog No. 50-000-79029 Supplier Meso Scale Discovery Supplier No. K1519PR2
The Meso Scale Discovery Human Abeta38 6E10 Kit is a multiplex immunoassay that quantitatively measures amyloid beta 38 (Abeta38) in human samples such as serum plasma CSF and cell culture supernatants This electrochemiluminescence (ECL) assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium calibrator and sample diluent The 6E10 antibody enables specific detection of Abeta38 This assay allows high-throughput quantification of Abeta38 levels which may be useful as a biomarker in Alzheimer s disease research