Learn More
Meso Scale Discovery R-PLEX Human AB38 6E10 ASYQP
SDP
Catalog No. 5000079447
Encompass
The R-PLEX Human AB38 6E10 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures A 38 and A 40 in human samples such as CSF serum and plasma This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ECL tags calibrators and controls This assay allows simultaneous measurement of A 38 and A 40 and is suitable for research use only
Catalog No. 50-000-79447 Supplier Meso Scale Discovery Supplier No. K1519PR22
The R-PLEX Human AB38 6E10 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures A 38 and A 40 in human samples such as CSF serum and plasma This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ECL tags calibrators and controls This assay allows simultaneous measurement of A 38 and A 40 and is suitable for research use only