Learn More
Meso Scale Discovery R-PLEX Human AB40 6E10 ASY
SDP
Catalog No. 5000080630
Encompass
The Meso Scale Discovery R-PLEX Human Abeta40 6E10 Assay is a multiplex immunoassay kit that allows simultaneous detection of amyloid beta 40 (Abeta40) and amyloid beta bound by antibody 6E10 in human samples such as serum plasma and CSF It utilizes electrochemiluminescence (ECL) detection technology on patterned arrays for high sensitivity and a broad dynamic range The assay requires 50 L or less of sample and can be run in as little as 3 5 hours It includes a plate coated with capture antibodies detection antibodies labeled with ECL tags calibrators controls 10X diluent 4X read buffer and wash buffer
Catalog No. 50-000-80630 Supplier Meso Scale Discovery Supplier No. K1518RR2
Add to Cart
Add to Cart
The Meso Scale Discovery R-PLEX Human Abeta40 6E10 Assay is a multiplex immunoassay kit that allows simultaneous detection of amyloid beta 40 (Abeta40) and amyloid beta bound by antibody 6E10 in human samples such as serum plasma and CSF It utilizes electrochemiluminescence (ECL) detection technology on patterned arrays for high sensitivity and a broad dynamic range The assay requires 50 L or less of sample and can be run in as little as 3 5 hours It includes a plate coated with capture antibodies detection antibodies labeled with ECL tags calibrators controls 10X diluent 4X read buffer and wash buffer