Learn More
Meso Scale Discovery R-PLEX Human AB40 6E10 ASYQP
SDP
Catalog No. 5000078415
Encompass
The Meso Scale Discovery R-PLEX Human Abeta40 6E10 Assay is a multiplex immunoassay kit that allows simultaneous measurement of amyloid beta 40 (Abeta40) and total amyloid beta (6E10) in human samples such as CSF serum and plasma It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L of sample and can be read on MSD instrument platforms It allows quantification of Abeta40 and Abeta peptides which are biomarkers associated with Alzheimer s disease This multiplexed assay enables efficient and accurate analysis of key Alzheimer s biomarkers
Catalog No. 50-000-78415 Supplier Meso Scale Discovery Supplier No. K1518RR22
The Meso Scale Discovery R-PLEX Human Abeta40 6E10 Assay is a multiplex immunoassay kit that allows simultaneous measurement of amyloid beta 40 (Abeta40) and total amyloid beta (6E10) in human samples such as CSF serum and plasma It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L of sample and can be read on MSD instrument platforms It allows quantification of Abeta40 and Abeta peptides which are biomarkers associated with Alzheimer s disease This multiplexed assay enables efficient and accurate analysis of key Alzheimer s biomarkers