Learn More
Meso Scale Discovery R-PLEX Human AB42 6E10 ASY
SDP
Catalog No. 5000080156
Encompass
The Meso Scale Discovery R-Plex Human A 42/A 40 Assay Kit is a multiplex immunoassay that measures A 42 and A 40 in human samples such as CSF serum and plasma It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample volume requirements The assay has a testing time of 3 5 hours and shows no cross-reactivity with other amyloid proteins It requires a Meso Scale Discovery instrument such as QuickPlex QuickPlex SQ 120 or QuickPlex SQ 240 to run the assay This product enables the reliable measurement of A 42 and A 40 levels which aids in Alzheimer s disease research and drug development
Catalog No. 50-000-80156 Supplier Meso Scale Discovery Supplier No. K1518SR2
The Meso Scale Discovery R-Plex Human A 42/A 40 Assay Kit is a multiplex immunoassay that measures A 42 and A 40 in human samples such as CSF serum and plasma It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample volume requirements The assay has a testing time of 3 5 hours and shows no cross-reactivity with other amyloid proteins It requires a Meso Scale Discovery instrument such as QuickPlex QuickPlex SQ 120 or QuickPlex SQ 240 to run the assay This product enables the reliable measurement of A 42 and A 40 levels which aids in Alzheimer s disease research and drug development