Learn More
Meso Scale Discovery R-PLEX Human ACE ASY
SDP
Catalog No. 5000078658
Encompass
The R-PLEX Human ACE Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures angiotensin converting enzyme (ACE) levels in human serum plasma urine cell culture supernatants and other biological samples The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay can quantify ACE levels in approximately 3 5 hours
Catalog No. 50-000-78658 Supplier Meso Scale Discovery Supplier No. K151J8R2
Add to Cart
Add to Cart
The R-PLEX Human ACE Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures angiotensin converting enzyme (ACE) levels in human serum plasma urine cell culture supernatants and other biological samples The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay can quantify ACE levels in approximately 3 5 hours