Learn More
Meso Scale Discovery R-PLEX Human ACE ASYQP
SDP
Catalog No. 5000078659
Encompass
The R-PLEX Human ACE Assay Kit from Meso Scale Discovery is used to quantitatively measure angiotensin converting enzyme (ACE) levels in human serum plasma urine cell culture supernatants and tissue homogenates The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volumes The kit includes a 96-well plate coated with a monoclonal antibody to human ACE detection antibody read buffer wash buffer blocker solutions calibrator and positive control The assay can be read on MSD instrument platforms and takes approximately 4 hours to complete
Catalog No. 50-000-78659 Supplier Meso Scale Discovery Supplier No. K151J8R22
The R-PLEX Human ACE Assay Kit from Meso Scale Discovery is used to quantitatively measure angiotensin converting enzyme (ACE) levels in human serum plasma urine cell culture supernatants and tissue homogenates The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volumes The kit includes a 96-well plate coated with a monoclonal antibody to human ACE detection antibody read buffer wash buffer blocker solutions calibrator and positive control The assay can be read on MSD instrument platforms and takes approximately 4 hours to complete