Learn More
Meso Scale Discovery R-PLEX Human ACE2 ASY
SDP
Catalog No. 5000081617
Encompass
The R-PLEX Human ACE2 Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures human angiotensin-converting enzyme 2 (ACE2) levels in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can help monitor ACE2 levels which may be relevant in COVID-19 infection and research This 10-plex assay includes a total of 10 analytes including human ACE2 and 9 other biomarkers
Catalog No. 50-000-81617 Supplier Meso Scale Discovery Supplier No. K151AHUR2
The R-PLEX Human ACE2 Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures human angiotensin-converting enzyme 2 (ACE2) levels in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can help monitor ACE2 levels which may be relevant in COVID-19 infection and research This 10-plex assay includes a total of 10 analytes including human ACE2 and 9 other biomarkers