Learn More
Meso Scale Discovery R-PLEX Human ACE2 ASYQP
SDP
Catalog No. 5000081784
Encompass
The R-PLEX Human ACE2 Assay from Meso Scale Discovery measures human ACE2 levels in serum plasma and cell culture supernatants using MSD s electrochemiluminescence technology It offers high sensitivity a dynamic range of up to 5 logs and low sample volume requirements The assay quantifies ACE2 levels from 0 05 to 10 000 pg/mL with high specificity and minimal cross-reactivity Available as a singleplex assay it includes a plate with capture antibody detection antibody and calibrator aiding in the study of ACE2 levels related to COVID-19 infection and progression
Catalog No. 50-000-81784 Supplier Meso Scale Discovery Supplier No. K151AHUR22
The R-PLEX Human ACE2 Assay from Meso Scale Discovery measures human ACE2 levels in serum plasma and cell culture supernatants using MSD s electrochemiluminescence technology It offers high sensitivity a dynamic range of up to 5 logs and low sample volume requirements The assay quantifies ACE2 levels from 0 05 to 10 000 pg/mL with high specificity and minimal cross-reactivity Available as a singleplex assay it includes a plate with capture antibody detection antibody and calibrator aiding in the study of ACE2 levels related to COVID-19 infection and progression