Learn More
Meso Scale Discovery R-PLEX Human ADAM 9 ASY
SDP
Catalog No. 5000079873
Encompass
The R-PLEX Human ADAM-9 Assay is an ELISA kit for measuring ADAM-9 in human serum plasma and cell culture supernatants It uses a sandwich enzyme immunoassay with a monoclonal antibody for ADAM-9 on a microplate Samples bind to the antibody and after washing an enzyme-linked polyclonal antibody is added A substrate solution produces a color change proportional to ADAM-9 levels which is measured against a standard curve
Catalog No. 50-000-79873 Supplier Meso Scale Discovery Supplier No. K151ABHR2
The R-PLEX Human ADAM-9 Assay is an ELISA kit for measuring ADAM-9 in human serum plasma and cell culture supernatants It uses a sandwich enzyme immunoassay with a monoclonal antibody for ADAM-9 on a microplate Samples bind to the antibody and after washing an enzyme-linked polyclonal antibody is added A substrate solution produces a color change proportional to ADAM-9 levels which is measured against a standard curve