Learn More
Meso Scale Discovery R-PLEX Human ADAM-TS 13 ASY
SDP
Catalog No. 5000079481
Encompass
The R-PLEX Human ADAM-TS13 Assay Kit from Meso Scale Discovery measures ADAM-TS13 in human serum plasma and other samples using MSD s electrochemiluminescent detection It offers high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibodies detection antibody solution read buffer wash buffer diluent and calibrators for a standard curve Detection limits range from 50 to 100 000 pg/mL This kit accurately quantifies ADAM-TS13 crucial for blood coagulation regulation
Catalog No. 50-000-79481 Supplier Meso Scale Discovery Supplier No. K151AABR2
The R-PLEX Human ADAM-TS13 Assay Kit from Meso Scale Discovery measures ADAM-TS13 in human serum plasma and other samples using MSD s electrochemiluminescent detection It offers high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibodies detection antibody solution read buffer wash buffer diluent and calibrators for a standard curve Detection limits range from 50 to 100 000 pg/mL This kit accurately quantifies ADAM-TS13 crucial for blood coagulation regulation