Learn More
Meso Scale Discovery R-PLEX Human ADAM17 ASY
SDP
Catalog No. 5000081948
Encompass
The R-PLEX Human ADAM17 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures active ADAM17 in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibody a detection antibody and calibrator The assay can detect ADAM17 levels between 1 5-1000 pg/mL and each kit provides sufficient reagents for 1 x 96-well plate This product enables the accurate quantitation of ADAM17 which plays a key role in regulating EGFR signaling
Catalog No. 50-000-81948 Supplier Meso Scale Discovery Supplier No. K151ALWR2
The R-PLEX Human ADAM17 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures active ADAM17 in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibody a detection antibody and calibrator The assay can detect ADAM17 levels between 1 5-1000 pg/mL and each kit provides sufficient reagents for 1 x 96-well plate This product enables the accurate quantitation of ADAM17 which plays a key role in regulating EGFR signaling