Learn More
Meso Scale Discovery R-PLEX Human ADAM17 ASYQP
SDP
Catalog No. 5000078327
Encompass
The R-PLEX Human ADAM17 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures active ADAM17 in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity and a broad dynamic range This kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator The assay can detect ADAM17 levels between 0 3-1000 pg/mL and requires only 50 L of sample
Catalog No. 50-000-78327 Supplier Meso Scale Discovery Supplier No. K151ALWR22
The R-PLEX Human ADAM17 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures active ADAM17 in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity and a broad dynamic range This kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer and calibrator The assay can detect ADAM17 levels between 0 3-1000 pg/mL and requires only 50 L of sample