Learn More
Meso Scale Discovery R-PLEX Human AFP ASY
SDP
Catalog No. 5000079588
Encompass
The R-PLEX Human AFP Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of alpha-fetoprotein (AFP) in human serum plasma and tissue culture supernatant This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide a highly sensitive and reproducible method for detecting AFP an important biomarker used to help diagnose and monitor hepatocellular carcinoma and other malignancies as well as fetal defects The assay has a wide dynamic range of 1 95-2000 pg/mL and requires only 50 L of sample
Catalog No. 50-000-79588 Supplier Meso Scale Discovery Supplier No. K151AACR2
The R-PLEX Human AFP Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of alpha-fetoprotein (AFP) in human serum plasma and tissue culture supernatant This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide a highly sensitive and reproducible method for detecting AFP an important biomarker used to help diagnose and monitor hepatocellular carcinoma and other malignancies as well as fetal defects The assay has a wide dynamic range of 1 95-2000 pg/mL and requires only 50 L of sample