Learn More
Meso Scale Discovery R-PLEX Human AFP ASYQP
SDP
Catalog No. 5000079619
Encompass
The R-PLEX Human AFP Assay is an immunoassay panel that quantitatively measures alpha-fetoprotein (AFP) in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescence (ECL) on the Meso Scale Discovery (MSD) platform providing high sensitivity and a broad dynamic range This multiplex assay allows researchers to measure AFP and other biomarkers simultaneously from a small sample volume It has been validated for use in biomarker research related to hepatocellular carcinoma and other AFP-associated diseases The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required to perform the assay
Catalog No. 50-000-79619 Supplier Meso Scale Discovery Supplier No. K151AACR22
The R-PLEX Human AFP Assay is an immunoassay panel that quantitatively measures alpha-fetoprotein (AFP) in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescence (ECL) on the Meso Scale Discovery (MSD) platform providing high sensitivity and a broad dynamic range This multiplex assay allows researchers to measure AFP and other biomarkers simultaneously from a small sample volume It has been validated for use in biomarker research related to hepatocellular carcinoma and other AFP-associated diseases The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents required to perform the assay