Learn More
Meso Scale Discovery R-PLEX Human Aiolos ASYQP
SDP
Catalog No. 5000078883
Encompass
The R-PLEX Human AIolos Assay from Meso Scale Diagnostics is a multiplex immunoassay kit that enables simultaneous quantification of the AIolos protein in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows for efficient and reproducible analysis of AIolos levels in research studies
Catalog No. 50-000-78883 Supplier Meso Scale Discovery Supplier No. K1516TR22
The R-PLEX Human AIolos Assay from Meso Scale Diagnostics is a multiplex immunoassay kit that enables simultaneous quantification of the AIolos protein in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows for efficient and reproducible analysis of AIolos levels in research studies