Learn More
Meso Scale Discovery R-PLEX Human Albumin ASYQP
SDP
Catalog No. 5000080557
Encompass
The R-PLEX Human Albumin Assay Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of human albumin in biological samples such as serum plasma urine and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform testing The assay has a typical sensitivity of 1 g/mL and an upper quantitation limit of 300 g/mL albumin
Catalog No. 50-000-80557 Supplier Meso Scale Discovery Supplier No. K1514VR22
Add to Cart
Add to Cart
The R-PLEX Human Albumin Assay Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of human albumin in biological samples such as serum plasma urine and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform testing The assay has a typical sensitivity of 1 g/mL and an upper quantitation limit of 300 g/mL albumin