Learn More
Meso Scale Discovery R-PLEX Human ALK ASY
SDP
Catalog No. 5000078465
Encompass
The R-PLEX Human ALK Assay QuickPlex 5-PL Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring ALK in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection on the QuickPlex SQ 120 instrument The assay takes about 3 hours and needs 12 5 L of sample with a detection range of 19 to 10 000 pg/mL The kit includes reagents for calibration controls diluent detection antibodies read buffer and plates with capture antibodies
Catalog No. 50-000-78465 Supplier Meso Scale Discovery Supplier No. K151ALUR2
The R-PLEX Human ALK Assay QuickPlex 5-PL Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring ALK in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection on the QuickPlex SQ 120 instrument The assay takes about 3 hours and needs 12 5 L of sample with a detection range of 19 to 10 000 pg/mL The kit includes reagents for calibration controls diluent detection antibodies read buffer and plates with capture antibodies