Learn More
Meso Scale Discovery R-PLEX Human ALK ASYQP
SDP
Catalog No. 5000081947
Encompass
The R-PLEX Human ALK Assay QuickPlex 5-PL Kit from Meso Scale Diagnostics is a multiplex immunoassay for quantifying human Anaplastic Lymphoma Kinase (ALK) in cell lysates and other samples It uses electrochemiluminescence detection and includes reagents for 5-plex analysis of ALK and 4 other analytes The kit features a 96-well plate with capture antibodies detection antibodies calibrators controls buffers and seals It has a sensitivity of 0 3 ng/mL and a 3 5-hour assay time ideal for ALK level and activation state research
Catalog No. 50-000-81947 Supplier Meso Scale Discovery Supplier No. K151ALUR22
Add to Cart
Add to Cart
The R-PLEX Human ALK Assay QuickPlex 5-PL Kit from Meso Scale Diagnostics is a multiplex immunoassay for quantifying human Anaplastic Lymphoma Kinase (ALK) in cell lysates and other samples It uses electrochemiluminescence detection and includes reagents for 5-plex analysis of ALK and 4 other analytes The kit features a 96-well plate with capture antibodies detection antibodies calibrators controls buffers and seals It has a sensitivity of 0 3 ng/mL and a 3 5-hour assay time ideal for ALK level and activation state research