Learn More
Meso Scale Discovery R-PLEX Human AMICA1 ASYQP
SDP
Catalog No. 5000078855
Encompass
The R-PLEX Human AMICA1 Assay is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of AMICA1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and is compatible with the SECTOR Imager 2400A from Meso Scale Discovery The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 2500 pg/mL It includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls The total assay time is 3 5 hours This product can aid in understanding AMICA1 biology and evaluating AMICA1 as a biomarker
Catalog No. 50-000-78855 Supplier Meso Scale Discovery Supplier No. K151ABJR22
The R-PLEX Human AMICA1 Assay is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of AMICA1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and is compatible with the SECTOR Imager 2400A from Meso Scale Discovery The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 2500 pg/mL It includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls The total assay time is 3 5 hours This product can aid in understanding AMICA1 biology and evaluating AMICA1 as a biomarker