Learn More
Meso Scale Discovery R-PLEX Human Ang-1 ASY
SDP
Catalog No. 5000081803
Encompass
The R-PLEX Human Ang-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Angiopoietin-1 levels in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology allowing simultaneous quantitative measurement of Ang-1 and other biomarkers The assay has high sensitivity a broad dynamic range and requires low sample volumes This product provides a rapid reproducible method for measuring Ang-1 which plays a role in vascular development and angiogenesis
Catalog No. 50-000-81803 Supplier Meso Scale Discovery Supplier No. K151YQR2
The R-PLEX Human Ang-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures Angiopoietin-1 levels in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology allowing simultaneous quantitative measurement of Ang-1 and other biomarkers The assay has high sensitivity a broad dynamic range and requires low sample volumes This product provides a rapid reproducible method for measuring Ang-1 which plays a role in vascular development and angiogenesis