Learn More
Meso Scale Discovery R-PLEX Human Ang-4 ASY
SDP
Catalog No. 5000079713
Encompass
The R-PLEX Human ANG-4 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring Angiopoietin-4 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for simultaneous measurement of ANG-4 and other biomarkers The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents Results are available in 3 5 hours with high sensitivity and a dynamic range from 0 64 pg/mL to 10 000 pg/mL aiding in profiling ANG-4 and related proteins
Catalog No. 50-000-79713 Supplier Meso Scale Discovery Supplier No. K151Y3R2
The R-PLEX Human ANG-4 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring Angiopoietin-4 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for simultaneous measurement of ANG-4 and other biomarkers The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents Results are available in 3 5 hours with high sensitivity and a dynamic range from 0 64 pg/mL to 10 000 pg/mL aiding in profiling ANG-4 and related proteins