Learn More
Meso Scale Discovery R-PLEX Human Ang-like 3 ASY
SDP
Catalog No. 5000079597
Encompass
The R-PLEX Human Ang-like 3 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring Ang-like 3 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents Ang-like 3 regulates blood pressure lipid and glucose metabolism and insulin sensitivity aiding research on metabolic disorders
Catalog No. 50-000-79597 Supplier Meso Scale Discovery Supplier No. K151X3R2
The R-PLEX Human Ang-like 3 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring Ang-like 3 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and necessary reagents Ang-like 3 regulates blood pressure lipid and glucose metabolism and insulin sensitivity aiding research on metabolic disorders