Learn More
Meso Scale Discovery R-PLEX Human Ang-like 3 ASYQP
SDP
Catalog No. 5000081992
Encompass
The R-PLEX Human Ang-like 3 Assay QuickPlex Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for the quantitative measurement of Ang-like 3 in human serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence detection technology and provides reagents for the quantitative measurement of Ang-like 3 on the QuickPlex SQ 120 instrument The assay has a reportable range of 0 06 - 1000 ng/mL and requires 25 L sample The total assay time is approximately 2 5 hours This product provides a rapid and sensitive immunoassay for measuring Ang-like 3 levels in human samples
Catalog No. 50-000-81992 Supplier Meso Scale Discovery Supplier No. K151X3R22
The R-PLEX Human Ang-like 3 Assay QuickPlex Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for the quantitative measurement of Ang-like 3 in human serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence detection technology and provides reagents for the quantitative measurement of Ang-like 3 on the QuickPlex SQ 120 instrument The assay has a reportable range of 0 06 - 1000 ng/mL and requires 25 L sample The total assay time is approximately 2 5 hours This product provides a rapid and sensitive immunoassay for measuring Ang-like 3 levels in human samples