Learn More
Meso Scale Discovery R-PLEX Human Ang-like 4 ASY
SDP
Catalog No. 5000080482
Encompass
The R-PLEX Human Ang-like 4 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring Ang-like 4 in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture and detection antibodies buffers diluent and calibrators Sensitivity is 0 08 ng/mL with a quantification limit of 2500 ng/mL This assay enables high-throughput profiling of Ang-like 4 linked to lipid metabolism
Catalog No. 50-000-80482 Supplier Meso Scale Discovery Supplier No. K151Z3R2
The R-PLEX Human Ang-like 4 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring Ang-like 4 in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture and detection antibodies buffers diluent and calibrators Sensitivity is 0 08 ng/mL with a quantification limit of 2500 ng/mL This assay enables high-throughput profiling of Ang-like 4 linked to lipid metabolism