Learn More
Meso Scale Discovery R-PLEX Human Ang-like 6 ASY
SDP
Catalog No. 5000078960
Encompass
The R-PLEX Human Ang-like 6 Assay is a multiplex immunoassay kit for measuring Ang-like 6 protein in human serum plasma and cell culture supernatants It uses a quantitative sandwich enzyme immunoassay with color-coded magnetic beads pre-coated with specific antibodies The kit includes a 96-well plate standard proteins detection antibodies streptavidin-PE diluents wash buffer substrate stop solution and beads It detects Ang-like 6 concentrations from 31 3-2000 pg/mL with high sensitivity and specificity facilitating high-throughput screening and understanding of Ang-like 6 s role in biological processes and diseases
Catalog No. 50-000-78960 Supplier Meso Scale Discovery Supplier No. K151A4R2
The R-PLEX Human Ang-like 6 Assay is a multiplex immunoassay kit for measuring Ang-like 6 protein in human serum plasma and cell culture supernatants It uses a quantitative sandwich enzyme immunoassay with color-coded magnetic beads pre-coated with specific antibodies The kit includes a 96-well plate standard proteins detection antibodies streptavidin-PE diluents wash buffer substrate stop solution and beads It detects Ang-like 6 concentrations from 31 3-2000 pg/mL with high sensitivity and specificity facilitating high-throughput screening and understanding of Ang-like 6 s role in biological processes and diseases