Learn More
Meso Scale Discovery R-PLEX Human Ang-like 6 ASYQP
SDP
Catalog No. 5000079432
Encompass
The R-PLEX Human ANG-like 6 Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring Angiopoietin-like 6 (ANGPTL6) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity up to 7 logs of dynamic range and low sample volume needs The assay detects ANGPTL6 as low as 61 pg/mL with just 25 L of sample The kit includes a 96-well plate with capture antibody detection antibody read buffer calibration standard and sample diluent for accurate ANGPTL6 quantification in research
Catalog No. 50-000-79432 Supplier Meso Scale Discovery Supplier No. K151A4R22
The R-PLEX Human ANG-like 6 Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring Angiopoietin-like 6 (ANGPTL6) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity up to 7 logs of dynamic range and low sample volume needs The assay detects ANGPTL6 as low as 61 pg/mL with just 25 L of sample The kit includes a 96-well plate with capture antibody detection antibody read buffer calibration standard and sample diluent for accurate ANGPTL6 quantification in research