Learn More
Meso Scale Discovery R-PLEX Human Angiogenin ASY
SDP
Catalog No. 5000080483
Encompass
The R-PLEX Human Angiogenin Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures angiogenin levels in human serum plasma and other biological samples This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents needed to run the assay Results are obtained in as little as 3 5 hours This assay can be used in biomarker research related to cancer inflammation angiogenesis and other diseases
Catalog No. 50-000-80483 Supplier Meso Scale Discovery Supplier No. K151Z4R2
The R-PLEX Human Angiogenin Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures angiogenin levels in human serum plasma and other biological samples This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents needed to run the assay Results are obtained in as little as 3 5 hours This assay can be used in biomarker research related to cancer inflammation angiogenesis and other diseases