Learn More
Meso Scale Discovery R-PLEX Human Angiogenin ASYQP
SDP
Catalog No. 5000078782
Encompass
The R-PLEX Human Angiogenin Assay Kit from Meso Scale Discovery is used to quantitatively measure levels of human angiogenin in serum plasma cell culture supernatants cell and tissue lysates It utilizes an electrochemiluminescent detection method and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L of sample per well and takes approximately 4 hours to complete It includes a 96-well plate coated with capture antibody detection antibody Read Buffer and angiogenin calibrator This product can aid in research related to angiogenesis cancer inflammation and wound healing
Catalog No. 50-000-78782 Supplier Meso Scale Discovery Supplier No. K151Z4R22
Add to Cart
Add to Cart
The R-PLEX Human Angiogenin Assay Kit from Meso Scale Discovery is used to quantitatively measure levels of human angiogenin in serum plasma cell culture supernatants cell and tissue lysates It utilizes an electrochemiluminescent detection method and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L of sample per well and takes approximately 4 hours to complete It includes a 96-well plate coated with capture antibody detection antibody Read Buffer and angiogenin calibrator This product can aid in research related to angiogenesis cancer inflammation and wound healing