Learn More
Meso Scale Discovery R-PLEX Human Annexin A1 ASY
SDP
Catalog No. 5000079823
Encompass
The R-PLEX Human Annexin A1 Assay from Meso Scale Discovery is a multiplex immunoassay that measures Annexin A1 levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and diluents valid for serum EDTA plasma heparin plasma and cell culture supernatant samples The assay has a typical lower limit of detection of 0 04 ng/mL and an upper limit of quantification of 1500 ng/mL for Annexin A1
Catalog No. 50-000-79823 Supplier Meso Scale Discovery Supplier No. K151YKR2
The R-PLEX Human Annexin A1 Assay from Meso Scale Discovery is a multiplex immunoassay that measures Annexin A1 levels in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and diluents valid for serum EDTA plasma heparin plasma and cell culture supernatant samples The assay has a typical lower limit of detection of 0 04 ng/mL and an upper limit of quantification of 1500 ng/mL for Annexin A1