Learn More
Meso Scale Discovery R-PLEX Human Annexin A1 ASYQP
SDP
Catalog No. 5000079664
Encompass
The R-PLEX Human Annexin A1 Assay from Meso Scale Diagnostics is a multiplex immunoassay kit that allows for the quantitative measurement of Annexin A1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires an MSD instrument and provides components for one 96-well plate including coated plates calibrators controls diluent and detection antibodies It can detect Annexin A1 concentrations between 0 04 - 1500 ng/mL with an approximate 3 5 hour total assay time
Catalog No. 50-000-79664 Supplier Meso Scale Discovery Supplier No. K151YKR22
The R-PLEX Human Annexin A1 Assay from Meso Scale Diagnostics is a multiplex immunoassay kit that allows for the quantitative measurement of Annexin A1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires an MSD instrument and provides components for one 96-well plate including coated plates calibrators controls diluent and detection antibodies It can detect Annexin A1 concentrations between 0 04 - 1500 ng/mL with an approximate 3 5 hour total assay time