Learn More
Meso Scale Discovery R-PLEX Human Annexin A5 ASY
SDP
Catalog No. 5000079533
Encompass
The R-Plex Human Annexin A5 Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures Annexin A5 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay allows for the quantitative measurement of Annexin A5 an important biomarker for apoptosis It can be used to monitor cell death and apoptosis signaling in disease research drug discovery development and clinical research This multiplexed assay enables efficient and accurate analysis of Annexin A5 levels
Catalog No. 50-000-79533 Supplier Meso Scale Discovery Supplier No. K151AGER2
The R-Plex Human Annexin A5 Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures Annexin A5 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay allows for the quantitative measurement of Annexin A5 an important biomarker for apoptosis It can be used to monitor cell death and apoptosis signaling in disease research drug discovery development and clinical research This multiplexed assay enables efficient and accurate analysis of Annexin A5 levels